5-Chloro-3-[2-(5-chloro-2-methylthien-3-yl)cyclopent-1-en-1-yl]-2-methylthiophene 95%

Supplier: Ambeed
Warning

A992492-1G A992492-5G A992492-100MG A992492-250MG
76657-520EA 199 USD
76657-520 76657-522 77623-412 77623-414
5-Chloro-3-[2-(5-chloro-2-methylthien-3-yl)cyclopent-1-en-1-yl]-2-methylthiophene 95%
5-Chloro-3-[2-(5-chloro-2-methylthien-3-yl)cyclopent-1-en-1-yl]-2-methylthiophene

Precaución: for Research Use Only

Formula: C₁₅H₁₄Cl₂S₂
MDL Number: MFCD19382349
CAS Number: 219537-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR