tert-Butyl 5-bromo-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A992418-100MG A992418-1G A992418-250MG A992418-5G
76657-512EA 90.18 USD
76657-512 76657-514 76657-516 76657-518
tert-Butyl 5-bromo-2-methylbenzoate 95%
tert-Butyl 5-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
MW: 271.1503 g/mol
MDL Number: MFCD21508504
CAS Number: 1202551-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR