5,6-Dibromo-1H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Warning

A992335-1G A992335-5G
76657-488EA 198.39 USD
76657-488 76657-490
5,6-Dibromo-1H-benzo[d][1,2,3]triazole 95%
5,6-Dibromo-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₆H₃Br₂N₃
MW: 276.9161 g/mol
MDL Number: MFCD00795591
CAS Number: 716320-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR