2,5,8,11-Tetraoxatridecan-13-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: m-PEG3-CH2COOH

A992101-100MG A992101-250MG A992101-1G
76657-484EA 24.41 USD
76657-484 76657-486 77599-434
2,5,8,11-Tetraoxatridecan-13-oic acid 95%
2,5,8,11-Tetraoxatridecan-13-oic acid

Precaución: for Research Use Only

Formula: C₉H₁₈O₆
MW: 222.2356 g/mol
MDL Number: MFCD25959645
CAS Number: 16024-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR