(R)-2-((benzyloxy)methyl)morpholine 98%

Supplier: Ambeed
Warning

A992092-100MG A992092-1G A992092-250MG
76657-478EA 133.68 USD
76657-478 76657-480 76657-482
(R)-2-((benzyloxy)methyl)morpholine 98%
(R)-2-((benzyloxy)methyl)morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.2689 g/mol
MDL Number: MFCD24395785
CAS Number: 135065-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR