2-(3-(2-(4-Chlorophenyl)imidazo[1,2-a]pyridin-6-yl)phenyl)propan-2-ol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
NOT Receptor Modulator 1
A992031-100MG
A992031-250MG
76657-474EA
162
USD
76657-474
76657-476
2-(3-(2-(4-Chlorophenyl)imidazo[1,2-a]pyridin-6-yl)phenyl)propan-2-ol 95%
2-(3-(2-(4-Chlorophenyl)imidazo[1,2-a]pyridin-6-yl)phenyl)propan-2-ol
Precaución: for Research Use Only
Formula:
C₂₂H₁₉ClN₂O MW: 362.85 g/mol |
MDL Number:
MFCD31555261 CAS Number: 1015231-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...