4-Bromo-1,5-dimethylimidazole 95%

Supplier: Ambeed
Warning

A992009-100MG A992009-250MG A992009-1G
76657-470EA 118.83 USD
76657-470 76657-472 77595-604
4-Bromo-1,5-dimethylimidazole 95%
4-Bromo-1,5-dimethylimidazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂
MW: 175.026 g/mol
MDL Number: MFCD09033089
CAS Number: 158585-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR