Ethyl 6-methoxy-3-methylbenzofuran-2-carboxylate 95%

Supplier: Ambeed
Warning

A991992-100MG A991992-250MG
76657-466EA 12.74 USD
76657-466 76657-468
Ethyl 6-methoxy-3-methylbenzofuran-2-carboxylate 95%
Ethyl 6-methoxy-3-methylbenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₄
MW: 234.2479 g/mol
MDL Number: MFCD16658378
CAS Number: 53724-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR