2,4-Dichlorobenzo[4,5]thieno[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A991987-1G A991987-250MG A991987-25G A991987-5G
76657-458EA 10 USD
76657-458 76657-460 76657-462 76657-464
2,4-Dichlorobenzo[4,5]thieno[3,2-d]pyrimidine 95%
2,4-Dichlorobenzo[4,5]thieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₄Cl₂N₂S
MDL Number: MFCD13190347
CAS Number: 160199-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR