Propargyl methanesulfonate ester 97%

Supplier: Ambeed
Danger

A991976-1G A991976-5G
76657-454EA 9.55 USD
76657-454 76657-456
Propargyl methanesulfonate ester 97%
Propargyl methanesulfonate ester

Precaución: for Research Use Only

Formula: C₄H₆O₃S
MW: 134.1536 g/mol
MDL Number: MFCD09841205
CAS Number: 16156-58-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR