5-Amino-2-bromo-3-fluoropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Pyridinamine, 6-bromo-5-fluoro-, 6-bromo-5-fluoropyridin-3-amine

A991872-100MG A991872-250MG A991872-1G A991872-5G
76657-444EA 9.55 USD
76657-444 76657-446 77573-980 77595-602
5-Amino-2-bromo-3-fluoropyridine 95%
5-Amino-2-bromo-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MW: 191.00 g/mol
MDL Number: MFCD21099397
CAS Number: 1256276-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR