tert-Butyl hex-5-ynoate 98%

Supplier: Ambeed
Warning

A991728-100MG A991728-1G A991728-250MG A991728-5G
76657-432EA 64.72 USD
76657-432 76657-434 76657-436 77623-406
tert-Butyl hex-5-ynoate 98%
tert-Butyl hex-5-ynoate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
MW: 168.23 g/mol
MDL Number: MFCD13192076
CAS Number: 73448-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR