[3-(2-Propyn-1-ylcarbamoyl)phenyl]boronic acid 95%

Supplier: Ambeed
Warning

A991478-100MG A991478-1G A991478-250MG
76657-422EA 37.14 USD
76657-422 76657-424 76657-426
[3-(2-Propyn-1-ylcarbamoyl)phenyl]boronic acid 95%
[3-(2-Propyn-1-ylcarbamoyl)phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₃
MW: 203.0000 g/mol
MDL Number: MFCD04115703
CAS Number: 874459-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR