2-Bromo-1-(tert-butyl)-4-chlorobenzene 95%

Supplier: Ambeed
Warning

A991310-100MG A991310-1G A991310-250MG
76657-416EA 26.53 USD
76657-416 76657-418 76657-420
2-Bromo-1-(tert-butyl)-4-chlorobenzene 95%
2-Bromo-1-(tert-butyl)-4-chlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrCl
MW: 247.5593 g/mol
MDL Number: MFCD28356425
CAS Number: 1369776-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR