Methyl 3-cyano-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A991305-1G A991305-10G
76657-414EA 36.08 USD
76657-414 77573-976
Methyl 3-cyano-2-methylbenzoate 95%
Methyl 3-cyano-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.1840 g/mol
MDL Number: MFCD14584407
CAS Number: 93340-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR