(2-Chloropyrimidin-4-yl)methanol 95%

Supplier: Ambeed
Warning

A991272-100MG A991272-250MG
76657-408EA 85 USD
76657-408 76657-410
(2-Chloropyrimidin-4-yl)methanol 95%
(2-Chloropyrimidin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MDL Number: MFCD18449371
CAS Number: 34953-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR