Biphenyl-4,4'-diacetic acid 95%

Supplier: Ambeed
Warning

A991199-250MG A991199-50MG
76657-398EA 206 USD
76657-398 76657-400
Biphenyl-4,4'-diacetic acid 95%
Biphenyl-4,4'-diacetic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₄
MDL Number: MFCD09881113
CAS Number: 19806-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR