5-(Tetrahydrofuran-2-yl)-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A991060-250MG A991060-1G
76657-390EA 157 USD
76657-390 77784-668
5-(Tetrahydrofuran-2-yl)-1H-pyrazol-3-amine 95%
5-(Tetrahydrofuran-2-yl)-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O
MDL Number: MFCD29048688
CAS Number: 1028843-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR