Dibenzo[b,d]furan-4,6-diyldiboronic acid 97%

Supplier: Ambeed
Warning

A990895-250MG A990895-1G
76657-388EA 94.43 USD
76657-388 77623-394
Dibenzo[b,d]furan-4,6-diyldiboronic acid 97%
Dibenzo[b,d]furan-4,6-diyldiboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀B₂O₅
MW: 255.8268 g/mol
MDL Number: MFCD28016435
CAS Number: 145238-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR