2-(Benzyloxy)-1-methylpyridin-1-ium trifluoromethanesulfonate 96%

Supplier: Ambeed
Danger

A990879-1G A990879-25G A990879-250MG
76657-386EA 17 USD
76657-386 77569-724 77623-392
2-(Benzyloxy)-1-methylpyridin-1-ium trifluoromethanesulfonate 96%
2-(Benzyloxy)-1-methylpyridin-1-ium trifluoromethanesulfonate

Precaución: For research use only.

Formula: C₁₄H₁₄F₃NO₄S
MW: 349.3255 g/mol
MDL Number: MFCD09265129
CAS Number: 882980-43-0
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR