2-(4-Methylthiazol-5-yl)ethyl octanoate 98%

Supplier: Ambeed
Warning

A990729-25G A990729-5G
76657-372EA 22.28 USD
76657-372 76657-374
2-(4-Methylthiazol-5-yl)ethyl octanoate 98%
2-(4-Methylthiazol-5-yl)ethyl octanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₂S
MW: 269.4029 g/mol
MDL Number: MFCD11617929
CAS Number: 163266-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR