Nicotinamide riboside chloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium chloride

A990719-100G A990719-25G A990719-5G
76657-370EA 176.11 USD
76657-370 77552-828 77595-598
Nicotinamide riboside chloride 98%
Nicotinamide riboside chloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O₅
MW: 290.7002 g/mol
MDL Number: MFCD27952781
CAS Number: 23111-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR