(S)-3-Amino-piperidine-2,6-dione hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Aminopiperidine-2,6-dione hydrochloride

A990633-100MG A990633-1G A990633-250MG A990633-5G A990633-25G
76657-350EA 5 USD
76657-350 76657-352 76657-354 76657-356 77623-384
(S)-3-Amino-piperidine-2,6-dione hydrochloride 98%
(S)-3-Amino-piperidine-2,6-dione hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂O₂
MW: 164.5902 g/mol
MDL Number: MFCD15474943
CAS Number: 25181-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR