(2R,6S)-tert-butyl 2,6-dimethylmorpholine-4-carboxylate 95%

Supplier: Ambeed
Warning

A990238-1G A990238-25G
76657-338EA 12.74 USD
76657-338 76657-340
(2R,6S)-tert-butyl 2,6-dimethylmorpholine-4-carboxylate 95%
(2R,6S)-tert-butyl 2,6-dimethylmorpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.2893 g/mol
MDL Number: MFCD30284782
CAS Number: 1599378-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR