(3R,6R)-6-Methylpiperidine-3-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A990080-100MG
A990080-250MG
76657-324EA
253
USD
76657-324
76657-326
(3R,6R)-6-Methylpiperidine-3-carboxylic acid hydrochloride 95%
(3R,6R)-6-Methylpiperidine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD26743833 CAS Number: 2682097-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...