4,4',4''-[(Benzene-1,3,5-triyl)triethyne-2,1-diyl]tripyridine 97%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tris[(pyridin-4-yl)ethynyl]benzene, 1,3,5-Tris(pyridin-4-ylethynyl)benzene

A989903-100MG A989903-5G A989903-1G A989903-250MG
76657-320EA 46.68 USD
76657-320 76657-322 77595-588 77599-406
4,4',4''-[(Benzene-1,3,5-triyl)triethyne-2,1-diyl]tripyridine 97%
4,4',4''-[(Benzene-1,3,5-triyl)triethyne-2,1-diyl]tripyridine

Precaución: for Research Use Only

Formula: C₂₇H₁₅N₃
MW: 381.428 g/mol
MDL Number: MFCD25541760
CAS Number: 168289-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR