(E)-2-(5-Methoxy-2-nitrophenyl)-N,N-dimethylethenamine 90%

Supplier: Ambeed
Warning

A989855-100MG A989855-250MG
76657-310EA 56.23 USD
76657-310 76657-312
(E)-2-(5-Methoxy-2-nitrophenyl)-N,N-dimethylethenamine 90%
(E)-2-(5-Methoxy-2-nitrophenyl)-N,N-dimethylethenamine

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₃
MW: 222.2405 g/mol
MDL Number: MFCD27976478
CAS Number: 32989-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR