4-Bromo-1-((methylsulfonyl)methyl)-2-nitrobenzene 95%
Supplier: Ambeed
|
Warning
|
A989781-100MG
A989781-1G
A989781-250MG
76657-304EA
44.56
USD
76657-304
76657-306
76657-308
4-Bromo-1-((methylsulfonyl)methyl)-2-nitrobenzene 95%
4-Bromo-1-((methylsulfonyl)methyl)-2-nitrobenzene
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₄S MW: 294.1224 g/mol |
MDL Number:
MFCD27976473 CAS Number: 651780-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...