N-Boc-3-iodo-DL-phenylalanine 95%
Supplier: Ambeed
|
Warning
|
A989659-100MG
A989659-1G
A989659-250MG
76657-296EA
39
USD
76657-296
76657-298
76657-300
N-Boc-3-iodo-DL-phenylalanine 95%
N-Boc-3-iodo-DL-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₄H₁₈INO₄ |
MDL Number:
MFCD01860478 CAS Number: 273221-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...