6-(4-((4-Methoxyphenyl)diazenyl)phenoxy)hexyl methacrylate 95%

Supplier: Ambeed
Warning

A989643-1G A989643-250MG
76657-292EA 377 USD
76657-292 76657-294
6-(4-((4-Methoxyphenyl)diazenyl)phenoxy)hexyl methacrylate 95%
6-(4-((4-Methoxyphenyl)diazenyl)phenoxy)hexyl methacrylate

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₂O₄
MW: 396.4900 g/mol
MDL Number: MFCD31556221
CAS Number: 135529-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR