2,5-Dichloro-4-hydroxybenzonitrile 95%

Supplier: Ambeed
Danger

A989615-1G A989615-250MG A989615-5G
76657-280EA 69 USD
76657-280 76657-282 76657-284
2,5-Dichloro-4-hydroxybenzonitrile 95%
2,5-Dichloro-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO
MDL Number: MFCD22988323
CAS Number: 3336-18-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR