N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-phenyl-9H-fluoren-2-amine

A989538-1G A989538-250MG
76657-264EA 57.29 USD
76657-264 76657-266
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline 95%
N,N-Bis(9,9-dimethyl-9H-fluoren-2-yl)aniline

Precaución: for Research Use Only

Formula: C₃₆H₃₁N
MW: 477.65 g/mol
MDL Number: MFCD29089346
CAS Number: 165320-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR