2,4,6-Trimethoxybenzenethiol 97%

Supplier: Ambeed
Warning

A989502-100MG A989502-1G A989502-250MG
76657-260EA 67 USD
76657-260 76657-262 77784-650
2,4,6-Trimethoxybenzenethiol 97%
2,4,6-Trimethoxybenzenethiol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S
MDL Number: MFCD11189388
CAS Number: 77189-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR