1-Ethyl-2-fluoro-4-methoxy-5-nitrobenzene 98%

Supplier: Ambeed
Danger

A989398-250MG A989398-1G
76657-258EA 105 USD
76657-258 77599-392
1-Ethyl-2-fluoro-4-methoxy-5-nitrobenzene 98%
1-Ethyl-2-fluoro-4-methoxy-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₃
MDL Number: MFCD16658623
CAS Number: 1089282-52-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR