Di-tert-butyl carbonate 98%

Supplier: Ambeed
Warning

A989340-1G A989340-5G A989340-250MG
76657-254EA 24 USD
76657-254 76657-256 77623-362
Di-tert-butyl carbonate 98%
Di-tert-butyl carbonate

Precaución: for Research Use Only

Formula: C₉H₁₈O₃
MDL Number: MFCD14560560
CAS Number: 34619-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR