(4-Methylisoxazol-5-yl)methanol 95%

Supplier: Ambeed
Warning

A989148-100MG A989148-250MG
76657-240EA 131.56 USD
76657-240 76657-242
(4-Methylisoxazol-5-yl)methanol 95%
(4-Methylisoxazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₇NO₂
MW: 113.1200 g/mol
MDL Number: MFCD28549584
CAS Number: 1607024-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR