Cis-9-octadecenal 95%

Supplier: Ambeed
Warning

A989069-100MG A989069-250MG A989069-1G
76657-234EA 107 USD
76657-234 76657-236 77552-826
Cis-9-octadecenal 95%
Cis-9-octadecenal

Precaución: for Research Use Only

Formula: C₁₈H₃₄O
MDL Number: MFCD31556108
CAS Number: 2423-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR