6-[(tert-Butoxy)carbonyl]-6-azaspiro[2.5]octane-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A988988-100MG A988988-1G A988988-250MG A988988-50MG
76657-226EA 207.94 USD
76657-226 76657-228 76657-230 76657-232
6-[(tert-Butoxy)carbonyl]-6-azaspiro[2.5]octane-5-carboxylic acid 95%
6-[(tert-Butoxy)carbonyl]-6-azaspiro[2.5]octane-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄
MW: 255.3101 g/mol
MDL Number: MFCD30802932
CAS Number: 1417743-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR