2,3-Dichloro-4-fluorobenzonitrile 95%

Supplier: Ambeed
Warning

A988952-100MG A988952-1G A988952-250MG A988952-50MG
76657-218EA 21 USD
76657-218 76657-220 76657-222 76657-224
2,3-Dichloro-4-fluorobenzonitrile 95%
2,3-Dichloro-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂Cl₂FN
MDL Number: MFCD19690560
CAS Number: 908123-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR