Ethyl 5-fluoro-2-methylbenzoate 98+%

Supplier: Ambeed
Warning

A988946-100G A988946-25G A988946-5G
76657-212EA 147 USD
76657-212 76657-214 76657-216
Ethyl 5-fluoro-2-methylbenzoate 98+%
Ethyl 5-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD06204528
CAS Number: 56427-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR