2-Nitro-5-trifluoromethoxytoluene 95%

Supplier: Ambeed
Warning

A988804-10G A988804-1G A988804-250MG A988804-5G
76657-198EA 293 USD
76657-198 76657-200 76657-202 76657-204
2-Nitro-5-trifluoromethoxytoluene 95%
2-Nitro-5-trifluoromethoxytoluene

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MDL Number: MFCD18399927
CAS Number: 70692-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR