2-Methoxyphenyl trifluoromethanesulfonate 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Trifluoromethanesulphonic acid 2-Methoxyphenyl ester, 2-Methoxyphenyl triflate

A988714-1G A988714-25G A988714-5G
76657-182EA 32 USD
76657-182 76657-184 76657-186
2-Methoxyphenyl trifluoromethanesulfonate 98% (by GC)
2-Methoxyphenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₈H₇F₃O₄S
MW: 256.20 g/mol
MDL Number: MFCD25541795
CAS Number: 59099-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR