4'-Bromo-2',3'-difluoroacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-2,3-difluorophenyl)-ethanone

A988505-1G A988505-250MG
76657-174EA 135.8 USD
76657-174 76657-176
4'-Bromo-2',3'-difluoroacetophenone 95%
4'-Bromo-2',3'-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.0255 g/mol
MDL Number: MFCD13192197
CAS Number: 1007346-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR