2-Hydroxy-4-(4-methylthiazol-5-yl)benzonitrile 95%

Supplier: Ambeed
Warning

A988337-100MG A988337-1G A988337-250MG
76657-160EA 40.32 USD
76657-160 76657-162 76657-164
2-Hydroxy-4-(4-methylthiazol-5-yl)benzonitrile 95%
2-Hydroxy-4-(4-methylthiazol-5-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₈N₂OS
MDL Number: MFCD28965251
CAS Number: 1448190-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR