Benzyl aziridine-1-carboxylate 95%

Supplier: Ambeed
Warning

A987854-1G A987854-250MG
76657-144EA 327.82 USD
76657-144 76657-146
Benzyl aziridine-1-carboxylate 95%
Benzyl aziridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.1998 g/mol
MDL Number: MFCD16036962
CAS Number: 25711-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR