Methyl 6-cyano-5-hydroxypicolinate 90%

Supplier: Ambeed
Warning

A987744-100MG A987744-1G A987744-250MG
76657-136EA 82 USD
76657-136 76657-138 76657-140
Methyl 6-cyano-5-hydroxypicolinate 90%
Methyl 6-cyano-5-hydroxypicolinate

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MDL Number: MFCD18258396
CAS Number: 727736-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR