5-Chloro-2-fluoro-4-nitrophenol 97%

Supplier: Ambeed
Danger

A987730-100MG A987730-1G A987730-250MG
76657-124EA 5 USD
76657-124 76657-126 76657-128
5-Chloro-2-fluoro-4-nitrophenol 97%
5-Chloro-2-fluoro-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₃
MDL Number: MFCD28752304
CAS Number: 1500154-08-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR