(2-Oxo-1,3-dioxolan-4-yl)methyl methacrylate 95% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
Methacrylic acid (2-Oxo-1,3-dioxolan-4-yl)methyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A987700-1G
A987700-25G
A987700-5G
76657-122EA
54
USD
76657-122
77573-958
77623-324
(2-Oxo-1,3-dioxolan-4-yl)methyl methacrylate 95% stabilised
(2-Oxo-1,3-dioxolan-4-yl)methyl methacrylate
Precaución: for Research Use Only
Formula:
C₈H₁₀O₅ MW: 186.16 g/mol |
MDL Number:
MFCD28964174 CAS Number: 13818-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...