2-(Hexadecanoylamino)acetic acid 97%

Supplier: Ambeed
Warning

A987588-1G A987588-250MG A987588-5G
76657-108EA 35 USD
76657-108 76657-110 76657-112
2-(Hexadecanoylamino)acetic acid 97%
2-(Hexadecanoylamino)acetic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₅NO₃
MDL Number: MFCD00153471
CAS Number: 2441-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR