tert-Butyl 2-mercaptoacetate 95%

Supplier: Ambeed
Danger

A987171-10G A987171-1G A987171-250MG A987171-5G
76657-092EA 1229 USD
76657-092 76657-094 76657-096 76657-098
tert-Butyl 2-mercaptoacetate 95%
tert-Butyl 2-mercaptoacetate

Precaución: for Research Use Only

Formula: C₆H₁₂O₂S
MW: 148.2233 g/mol
MDL Number: MFCD25960778
CAS Number: 20291-99-0
UN: 3334
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR